Learn More
Electron Microscopy Sciences Globe White Glass Charged Slides, 90° Ground Edge, Corner, Blue Frosted Label
SDP
Supplier:  Electron Microscopy Sciences 7188384
Encompass_Preferred
Produced from the finest quality optical white glass, these charged microscope slides provide the ideal surface for cell and tissue adhesion. They are ideal for use in histology, cytology and pathology departments, where the convenience of an adhesive slide saves the user time and money. The slides are pre-cleaned and ready to use. Dimensions: 25 x 75 mm. Thickness: 1-1.2 mm
Edge: 90° Ground
Label: Blue Frosted
Catalog No. 50-192-8851