Learn More
Qosina Corp Open Jaw Slide Clamp | Green
Catalog No. NC1758541
Encompass
This clamp fits 0.197 (5mm) OD tubing and is made of polypropylene.
Catalog No. NC1758541 Supplier Qosina Corp Supplier No. 12053
This clamp fits 0.197 (5mm) OD tubing and is made of polypropylene.