Learn More
Electron Microscopy Sciences Slide Coating Adhesive 100 ML
SDP
Catalog No. 5028681
Encompass_Preferred
Prepared from 3-aminopropyltriethoxysilane in acetone. Good for coating glass slides to adhere paraffin sections for further processing without sections falling off from the slide. READY TO USE.
Catalog No. 50-286-81 Supplier Electron Microscopy Sciences Supplier No. 71302
Prepared from 3-aminopropyltriethoxysilane in acetone. Good for coating glass slides to adhere paraffin sections for further processing without sections falling off from the slide. READY TO USE.