Learn More
Electron Microscopy Sciences Microscope Slide Dispenser 1 each
SDP
Catalog No. 50930975
Encompass_Preferred
Molded of attractive gray ABA plastic. This dispenser provides a method of releasing clean, dust free 1"x3" slides, one at a time. Comes with a clear molded plastic cover to allow for visual checking of unused slides. Measurements: 4-3/8" x 4-7/8" x 5-1/2". Holds approximately 72 slides.
Catalog No. 50-930-975 Supplier Electron Microscopy Sciences Supplier No. 71300
Molded of attractive gray ABA plastic. This dispenser provides a method of releasing clean, dust free 1"x3" slides, one at a time. Comes with a clear molded plastic cover to allow for visual checking of unused slides. Measurements: 4-3/8" x 4-7/8" x 5-1/2". Holds approximately 72 slides.