Learn More
Mopec Slide Drying Hotplate
SDP
Catalog No. 22445329 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Microscope Slide Dryer
Catalog No. Product Type
22-445-329 Microscope Slide Dryer
1 options
Catalog No. 22-445-329 Supplier Mopec Supplier No. BK400
Add to Cart
Add to Cart
The Slide Drying Hotplate is designed to quickly and efficiently dry paraffin tissue sections. The rapid heating system ensures the hotplate is ready to use in less time, while the digital temperature control allows for accurate control of temperatures.
  • Designed to take up minimal bench-top space
  • Digital temperature control to meet precise temperature requirements
  • Rapid heating system ensures system is ready to use in less time
  • Slide support rails can be adjusted to fit different slide sizes
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Optional lid helps reduce drying time and improve temperature control
Specifications
Product Type Microscope Slide Dryer
Material Stainless Steel
Media Capacity 2
Temperature Range (Metric) 86°F to 194°F
Dimensions (L x W x H) 450 x 183 x 85.5 mm
Voltage 110/230 V