Learn More
Mopec™ Slide Drying Mini Workstation
SDP
Catalog No. 22445299 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Microscope Slide Dryer
Catalog No. Product Type
22-445-299 Microscope Slide Dryer
1 options
Catalog No. 22-445-299 Supplier Mopec™ Supplier No. BK225
Add to Cart
Add to Cart
This Slide Drying Mini Workstation for Flotation Baths was specifically designed to fit directly over the Illuminated Tissue Flotation Bath and take up minimal bench-top space.
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Mini hotplate removes wrinkles from tissue sections
  • Sloped drying area improves drying time and can hold up to 12 slides
  • Anti-microbial coating to help prevent bacteria and virus growth
Specifications
Product Type Microscope Slide Dryer
Slide Capacity 12