Learn More
Medicus Health Slide Drying Rack Holder, 15 Slides
SDP
Catalog No. NC1510992
Encompass
∙ 15 Place Slide Drying Rack cut from a solid block of polyethylene has many uses from hematology to histology ∙Slide Drying Racks are cut from a solid block of chemical-resistant polyethylene and are easy to clean∙Solid and strong with rubber feet for benchtop stability∙5528M1 - Holds 15 Slides - 152mm x 102mm x 21mm∙Use in Hematology to hold blood smear slides during drying∙Great for Microbiology to let Gram Stains dry∙Many uses in Histology and Cytology as well
Catalog No. NC1510992 Supplier Medicus Health Supplier No. 5528M1
∙ 15 Place Slide Drying Rack cut from a solid block of polyethylene has many uses from hematology to histology ∙Slide Drying Racks are cut from a solid block of chemical-resistant polyethylene and are easy to clean∙Solid and strong with rubber feet for benchtop stability∙5528M1 - Holds 15 Slides - 152mm x 102mm x 21mm∙Use in Hematology to hold blood smear slides during drying∙Great for Microbiology to let Gram Stains dry∙Many uses in Histology and Cytology as well