Learn More
Medicus Health Slide Drying Rack Holder, 40 Slides
SDP
Catalog No. NC1510993
Encompass
∙ 40 Place Slide Drying Rack cut from a solid block of polyethylene has many uses from hematology to histology ∙Slide Drying Racks are cut from a solid block of chemical-resistant polyethylene and are easy to clean∙Solid and strong with rubber feet for benchtop stability∙5528M2 - Holds 40 Slides - 298mm x 105mm x 24mm∙Use in Hematology to hold blood smear slides during drying∙Great for Microbiology to let Gram Stains dry∙Many uses in Histology and Cytology as well
Catalog No. NC1510993 Supplier Medicus Health Supplier No. 5528M2
Add to Cart
Add to Cart
∙ 40 Place Slide Drying Rack cut from a solid block of polyethylene has many uses from hematology to histology ∙Slide Drying Racks are cut from a solid block of chemical-resistant polyethylene and are easy to clean∙Solid and strong with rubber feet for benchtop stability∙5528M2 - Holds 40 Slides - 298mm x 105mm x 24mm∙Use in Hematology to hold blood smear slides during drying∙Great for Microbiology to let Gram Stains dry∙Many uses in Histology and Cytology as well