Learn More
StatLab Slide Drying Rack
SDP
Catalog No. 21033455
Change view
Click to view available options
No. of Racks:
50
Catalog No. No. of Racks
21-033-455 50
1 options
Catalog No. 21-033-455 Supplier StatLab Supplier No. SL670
Add to Cart
Add to Cart
Durable acrylic with 10 slots accommodate 50 slides
  • Acrylic slide rack, inclined, used for draining/drying slides
  • 10 slots accommodate 50 slides
  • 8.2 x 8.5 x 2.0 in. (front) x 4.45 in.(height) (rear)
Specifications
No. of Racks 50
Product Type Drying Rack