Learn More
Electron Microscopy Sciences Slide Drying Rack
SDP
Catalog No. NC1275951
Encompass_Preferred
The inclined acrylic slide rack is used for draining and drying microscope slides. It accommodates 50 microscope slides in 10 slots. Dimensions are 8.2 (W) x 8.5 (D) x 2.0 (front) x 4.45 (rear H) inches.
Catalog No. NC1275951 Supplier Electron Microscopy Sciences Supplier No. 70311
The inclined acrylic slide rack is used for draining and drying microscope slides. It accommodates 50 microscope slides in 10 slots. Dimensions are 8.2 (W) x 8.5 (D) x 2.0 (front) x 4.45 (rear H) inches.