Learn More
Electron Microscopy Sciences Slide Duplicating Mold, 3 Cavity
SDP
Catalog No. 5028682
Encompass_Preferred
Uses a 1x3" glass slide as a base, and duplicates the slide using the resin of your choice. Tissue sections are embedded against the flat glass surface. This duplicated resin slide can be studied under the Light Microscope, then cut, remounted, resectioned, and studied under the EM. A three cavity mold, each cavity measuring: 1"(W)x3"(L)x3/32"(D). Reference and Instructions enclosed
Catalog No. 50-286-82 Supplier Electron Microscopy Sciences Supplier No. 70170
Uses a 1x3" glass slide as a base, and duplicates the slide using the resin of your choice. Tissue sections are embedded against the flat glass surface. This duplicated resin slide can be studied under the Light Microscope, then cut, remounted, resectioned, and studied under the EM. A three cavity mold, each cavity measuring: 1"(W)x3"(L)x3/32"(D). Reference and Instructions enclosed