Learn More
Electron Microscopy Sciences Slide Duplicating withCover Slip
SDP
Catalog No. 5028683
Encompass_Preferred
The same design as above 70170 but with the addition of three round cavities which are 17 mm in diameter and 1 mm deep.
Catalog No. 50-286-83 Supplier Electron Microscopy Sciences Supplier No. 70172
The same design as above 70170 but with the addition of three round cavities which are 17 mm in diameter and 1 mm deep.