Learn More
Electron Microscopy Sciences SlideFile™ Junior Slide Storage, Yellow
SDP
Supplier:  Electron Microscopy Sciences 71448Y1
Encompass_Preferred
SlideFile Jr is a convenient, organized and versatile way of storing up to 200 slides in just 860cm3 (53 cu. in.). It is stackable to save space. Each SlideFile Jr. includes a slide box and a removable tray. A tinted hinged cover makes the contents of the box easy to see at a glance. Removable tray inside the storage box having 50 individual numbered slots. All slides are stored upright for easy insertion and removal as well as reading the bar codes without removing the slides. To save space , you can double the amount of the slides simply by storing 2 slides per slot. And for maximum storage space, simply remove the tray and line up 200 slides in 3 rows for long term storage. This system is not autoclavable. The system measures: 82 mm x 140 mm x 86 mm(H).
Color: Yellow
Catalog No. 50-931-080