Learn More
Electron Microscopy Sciences SlideFile™ Slide Storage System, Blue
SDP
Supplier:  Electron Microscopy Sciences 7145110
Encompass_Preferred
The most convenient, organized and versatile way of storing 3" x 1" (75 x 25 mm) microscope slides vertically. This impact resistant SlideFile Storage System can hold up to 400 slide per unit in just 1720 cm3 (105 cu. in.) and are stackable for space saving. Each SlideFile System includes a slide box and two removable trays. A transparent hinged cover makes it easy to see at a glance the content of the box.
Color: Blue
Catalog No. 50-189-9589