Learn More
Research Products International Corp Freezer Rack for 100 Place Slides, Stainless Steel, 3 x 6 Array, Dimensions (Inches) 9 x 22 x 9H
SDP
Catalog No. 50550272
Encompass
The slotted Microscope Slide Freezer Rack provides convenient storage for slide racks. Helps save time by keeping you organized in a cost efficient manner. The highest quality racks, constructed of corrosion resistant stainless steel, includes handles. Racks for 100 place slide boxes fit all 7 x 8 1/4 x 1 3/8 boxes.
Catalog No. 50-550-272 Supplier Research Products International Corp Supplier No. 247530
Add to Cart
Add to Cart
The slotted Microscope Slide Freezer Rack provides convenient storage for slide racks. Helps save time by keeping you organized in a cost efficient manner. The highest quality racks, constructed of corrosion resistant stainless steel, includes handles. Racks for 100 place slide boxes fit all 7 x 8 1/4 x 1 3/8 boxes.