Learn More
Med Vet International McKesson Frosted Microscope Slide, 1 x 3 Inch, 16-7132, 1440/Case
Catalog No. 503043156
Encompass_Preferred
Microscope Slide McKesson 1 X 3 Inch X 1 mm Frosted End Frosted 3 inches x 1 inch x 1 millimeter Economy slides designed to meet the needs of a variety of applications Ground edges
Catalog No. 50-304-3156 Supplier Med Vet International Supplier No. 464498CS
Microscope Slide McKesson 1 X 3 Inch X 1 mm Frosted End Frosted 3 inches x 1 inch x 1 millimeter Economy slides designed to meet the needs of a variety of applications Ground edges