Learn More
Med Vet International McKesson Premium Frosted Microscope Slide, 25 x 75 mm, 70-105PMCK, 1440/Case
Catalog No. 503045076
Encompass_Preferred
Microscope Slide McKesson 25 X 75 X 1 mm Frosted End 25 x 75 x 1 millimeters Frosted on one end Ground edges Made from the finest quality sheet glass Guaranteed precleaned Non-corrosive glass
Catalog No. 50-304-5076 Supplier Med Vet International Supplier No. 938362CS
Microscope Slide McKesson 25 X 75 X 1 mm Frosted End 25 x 75 x 1 millimeters Frosted on one end Ground edges Made from the finest quality sheet glass Guaranteed precleaned Non-corrosive glass