Learn More
Med Vet International McKesson Frosted Microscope Slide, 25 x 75 x 1 mm, 177-1380-50W, 72/Box
Catalog No. 503042943
Encompass_Preferred
Microscope Slide McKesson 25 X 75 X 1 mm White Frosted End Ground edge slide with 90° corners is perfect for use in automatic cover slip machines All are printed with McKesson under the color area for orientation purposes White, frosted end Pre-cleaned and ready to use 25 x 75 x 1 millimeters Six dozen microscope slides per box
Catalog No. 50-304-2943 Supplier Med Vet International Supplier No. 1082077BX
Microscope Slide McKesson 25 X 75 X 1 mm White Frosted End Ground edge slide with 90° corners is perfect for use in automatic cover slip machines All are printed with McKesson under the color area for orientation purposes White, frosted end Pre-cleaned and ready to use 25 x 75 x 1 millimeters Six dozen microscope slides per box