Learn More
Electron Microscopy Sciences Gold Seal Rite-On™ Slides, Frosted On One End One Side, 3 x 1", 1 mm thick
SDP
Supplier:  Electron Microscopy Sciences 6372105
Encompass_Preferred
GOLDSEAL slides are made from pure white glass (sodalime) and at accurately cut sizes. To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL slides are hermetically sealed in our Moisture-Barrier system, providing a dry-slide environments during shipment and storage.
Size: 3 x 1"
Thickness: 1 mm
Catalog No. 50-189-9121