Learn More
Electron Microscopy Sciences Globe Microscope Slides, Beveled Edge, Clipped Corner, Yellow Frosted Label
SDP
Catalog No. 501928843
Encompass_Preferred
Color frosted microscope slides with color coding on one end and one side. Pre-cleaned and ready to use. Edges specially angled at 45 degrees to provide secure grasp during blood smearing process.
Edge: Beveled
Corner: Clipped
Label: Yellow Frosted
Catalog No. 50-192-8843 Supplier Electron Microscopy Sciences Supplier No. 7188376
Color frosted microscope slides with color coding on one end and one side. Pre-cleaned and ready to use. Edges specially angled at 45 degrees to provide secure grasp during blood smearing process.
Edge: Beveled
Corner: Clipped
Label: Yellow Frosted