Learn More
Electron Microscopy Sciences Globe Slides Two-Sided Frosted, Ground Edge, Safety Corner
SDP
Catalog No. 501928795
Encompass_Preferred
These frosted microscope slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens.
Edge: Ground
Corner: Safety
Catalog No. 50-192-8795 Supplier Electron Microscopy Sciences Supplier No. 7188328
These frosted microscope slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens.
Edge: Ground
Corner: Safety