Learn More
Electron Microscopy Sciences Globe Diamond™ White Glass Frosted Slides One-Side, Beveled Edge, Clipped Corner
SDP
Supplier:  Electron Microscopy Sciences 7188308
Encompass_Preferred
Diamond White Glass frosted slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens. The slides are pre-cleaned and ready to use. Produced from the finest quality optical white glass.
Edge: Beveled
Corner: Clipped
Catalog No. 50-192-8775