Learn More
Electron Microscopy Sciences Globe Slides Two-Sided Frosted, Ground Edge, 90° Corner
SDP
Catalog No. 501928793
Encompass_Preferred
These frosted microscope slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens.
Edge: Ground
Corner: 90°
Catalog No. 50-192-8793 Supplier Electron Microscopy Sciences Supplier No. 7188326
These frosted microscope slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens.
Edge: Ground
Corner: 90°