Learn More
Electron Microscopy Sciences Globe Diamond™ White Glass Frosted Slides One-Side, Ground Edge, 90°Corner
SDP
Catalog No. 501928773
Encompass_Preferred
Diamond White Glass frosted slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens. The slides are pre-cleaned and ready to use. Produced from the finest quality optical white glass.
Edge: Ground
Corner: 90°
Catalog No. 50-192-8773 Supplier Electron Microscopy Sciences Supplier No. 7188306
Diamond White Glass frosted slides are produced using a unique chemical etching process. This provides the slides with a smooth frosted finish that won't wear out slide marking pens. The slides are pre-cleaned and ready to use. Produced from the finest quality optical white glass.
Edge: Ground
Corner: 90°