Learn More
Electron Microscopy Sciences Globe Microscope Slides, Ground Edge, 90° Corner, Green Frosted Label
SDP
Supplier:  Electron Microscopy Sciences 7188333
Encompass_Preferred
Color frosted microscope slides with color coding on one end and one side. Pre-cleaned and ready to use. Corners are 90 degree. Perfect for use in automatic cover slip machines.
Edge: Ground
Corner: 90°
Label: Green Frosted
Catalog No. 50-192-8800