Learn More
Electron Microscopy Sciences Globe Microscope Slides, Ground Edge, Safety Corner, Lilac Frosted Label
SDP
Supplier:  Electron Microscopy Sciences 7188353
Encompass_Preferred
Color frosted microscope slides with color coding on one end and one side. Pre-cleaned and ready to use. Slightly clipped corners for comfortable handling and preventing finger and hand cuts.
Edge: Ground
Corner: Safety
Label: Lilac Frosted
Catalog No. 50-192-8820