Learn More
Med Vet International Slide Holder Holds 25 Slides, Each
Catalog No. 502830198
Encompass_Preferred
Details: Sturdy ABS plastic box with our exclusive self-locking lid holds up to 25 slides and has cork lined bottom. Index paper in lid corresponds with embossed numerals on bottom of box.
Catalog No. 50-283-0198 Supplier Med Vet International Supplier No. SLIDEHOLDER
Details: Sturdy ABS plastic box with our exclusive self-locking lid holds up to 25 slides and has cork lined bottom. Index paper in lid corresponds with embossed numerals on bottom of box.