Learn More
Electron Microscopy Sciences Slide Holder
SDP
Catalog No. 501900415
Encompass_Preferred
Molded from polypropylene. Adjustable to hold 2 to 5 3x1" slides, or 10 slides back-to-back firmly. This Slide Holder works well with all Neat Stain Kits above.
Catalog No. 50-190-0415 Supplier Electron Microscopy Sciences Supplier No. 7141101
Molded from polypropylene. Adjustable to hold 2 to 5 3x1" slides, or 10 slides back-to-back firmly. This Slide Holder works well with all Neat Stain Kits above.