Learn More
Thomas Scientific Slide Holder for 78 Slides (polyethylene)
SDP
Catalog No. NC1223375
Encompass
Hold slides during evacuation, cleaning, and drying. Cut from chemical-resistant polyethylene. Solid and strong with rubber feet for benchtop stability.
Catalog No. NC1223375 Supplier Thomas Scientific Supplier No. 1216G61
Hold slides during evacuation, cleaning, and drying. Cut from chemical-resistant polyethylene. Solid and strong with rubber feet for benchtop stability.