Learn More
Andwin Scientific SLIDE HOLDERS
SDP
Catalog No. NC9418050
Encompass_Preferred
The Tissue-Tek Slide Holder, 24-place is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant.
Catalog No. NC9418050 Supplier Andwin Scientific Supplier No. 25608868
The Tissue-Tek Slide Holder, 24-place is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The product is solvent-resistant.