Learn More
Med Vet International Slide Holder, 25 Slides, Red, Each
Catalog No. 503045632
Encompass_Preferred
Sturdy ABS plastic box with our exclusive self locking lid holds up to 25 slides and has cork lined bottom. Index paper in lid corresponds with embossed numerals on bottom of box. Designed to stack. Packed 1 per box. Available options for this product: White | Blue | Green | Red | Yellow
Catalog No. 50-304-5632 Supplier Med Vet International Supplier No. SLIDEHOLDERREDEA
Sturdy ABS plastic box with our exclusive self locking lid holds up to 25 slides and has cork lined bottom. Index paper in lid corresponds with embossed numerals on bottom of box. Designed to stack. Packed 1 per box. Available options for this product: White | Blue | Green | Red | Yellow