Learn More
Ibidi USA µ-Slide III 3D Perfusion, Uncoated - Uncoated Polymer Coverslip
SDP
Catalog No. 501149025
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Slide III 3D Perfusion is an array of 6 wells where cells can be cultivated and, subsequently, investigated with microscopical methods. Two of the 6 wells respectively are connected by a channel. The channels can be connected to a pump for perfusing the wells in order to perform long term cell culture assays with 3D structures. #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box
This item is not returnable. View return policy
Catalog No. 50-114-9025 Supplier Ibidi USA Supplier No. 80371
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ–Slide III 3D Perfusion is an array of 6 wells where cells can be cultivated and, subsequently, investigated with microscopical methods. Two of the 6 wells respectively are connected by a channel. The channels can be connected to a pump for perfusing the wells in order to perform long term cell culture assays with 3D structures. #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box