Learn More
Electron Microscopy Sciences InkPro™ White Glass Microscope Slides, Blue
SDP
Catalog No. 501928585
Encompass_Preferred
InkPro Slides are made of white glass, specifically designed amd optimized for use with ink jet slide printing instruments. Print quality exhibits high resolution bar codes and alphanumerical characters for efficient and error-free scanning.
Color: Blue
Catalog No. 50-192-8585 Supplier Electron Microscopy Sciences Supplier No. 7186603
InkPro Slides are made of white glass, specifically designed amd optimized for use with ink jet slide printing instruments. Print quality exhibits high resolution bar codes and alphanumerical characters for efficient and error-free scanning.
Color: Blue