Learn More
Electron Microscopy Sciences InkPro™ White Glass Microscope Slides, Green
SDP
Catalog No. 501928586
Encompass_Preferred
InkPro Slides are made of white glass, specifically designed amd optimized for use with ink jet slide printing instruments. Print quality exhibits high resolution bar codes and alphanumerical characters for efficient and error-free scanning.
Color: Green
Catalog No. 50-192-8586 Supplier Electron Microscopy Sciences Supplier No. 7186604
InkPro Slides are made of white glass, specifically designed amd optimized for use with ink jet slide printing instruments. Print quality exhibits high resolution bar codes and alphanumerical characters for efficient and error-free scanning.
Color: Green