Learn More
Electron Microscopy Sciences InkPro™ White Glass Microscope Slides, White
SDP
Catalog No. 501928583
Encompass_Preferred
InkPro Slides are made of white glass, specifically designed amd optimized for use with ink jet slide printing instruments. Print quality exhibits high resolution bar codes and alphanumerical characters for efficient and error-free scanning.
Color: White
Catalog No. 50-192-8583 Supplier Electron Microscopy Sciences Supplier No. 7186601
InkPro Slides are made of white glass, specifically designed amd optimized for use with ink jet slide printing instruments. Print quality exhibits high resolution bar codes and alphanumerical characters for efficient and error-free scanning.
Color: White