Learn More
Electron Microscopy Sciences InkPro™ White Glass Microscope Slides, Yellow
SDP
Catalog No. 501928589
Encompass_Preferred
InkPro Slides are made of white glass, specifically designed amd optimized for use with ink jet slide printing instruments. Print quality exhibits high resolution bar codes and alphanumerical characters for efficient and error-free scanning.
Color: Yellow
Catalog No. 50-192-8589 Supplier Electron Microscopy Sciences Supplier No. 7186607
InkPro Slides are made of white glass, specifically designed amd optimized for use with ink jet slide printing instruments. Print quality exhibits high resolution bar codes and alphanumerical characters for efficient and error-free scanning.
Color: Yellow