Learn More
Electron Microscopy Sciences LockMailer™ Slide Jar Lockmailer
SDP
Catalog No. 5034092
Encompass_Preferred
This is a tamperproof multi purpose container for mailing, staining, or storing microscope slides. It is constructed of extra strong and opaque clear polypropylene, and high density polyethylene leakproof tamper evident color-coded screw cap. It will hold up to 4 standard 3 x 1" or 25 x 75 mm slides vertically. Inside chambers are slotted to keep slides safely separated. This LockMailer can be used as small staining vessel.
Catalog No. 50-340-92 Supplier Electron Microscopy Sciences Supplier No. 7140610
This is a tamperproof multi purpose container for mailing, staining, or storing microscope slides. It is constructed of extra strong and opaque clear polypropylene, and high density polyethylene leakproof tamper evident color-coded screw cap. It will hold up to 4 standard 3 x 1" or 25 x 75 mm slides vertically. Inside chambers are slotted to keep slides safely separated. This LockMailer can be used as small staining vessel.