Learn More
Newcomer Supply Ki-67 Control Slides, 10/set
SDP
Catalog No. NC0777904
Encompass
Newcomer Supply Ki-67 Control Slides are for the positive immunohistochemical staining of Ki-67, a marker of proliferating cell populations and useful as a prognostic indicator in a number of neoplasms. Ki-67 Control Slides contain sections of positive staining tonsil and negative staining adipose.
Catalog No. NC0777904 Supplier Newcomer Supply Supplier No. 3485A
Newcomer Supply Ki-67 Control Slides are for the positive immunohistochemical staining of Ki-67, a marker of proliferating cell populations and useful as a prognostic indicator in a number of neoplasms. Ki-67 Control Slides contain sections of positive staining tonsil and negative staining adipose.