Learn More
StatLab Slide Labels with Matte Finish
SDP
Catalog No. 23640013
Change view
Click to view available options
Type:
Slide Label
Catalog No. Type
23-640-013 Slide Label
1 options
Catalog No. 23-640-013 Supplier StatLab Supplier No. XRM1431A1CP
Add to Cart
Add to Cart
Xylene-resistant slide labels with matte finish, designed for reliable identification in laboratory environments.
  • Xylene-resistant slide labels measuring 0.9062 in x 7/8 in., designed to withstand harsh laboratory solvents and staining processes without smudging or degradation.
  • 1-across label format with a 1 in. core, perforated backing, matte finish, and opaque liner to support clear printing, easy handling, and dependable adhesion.
  • Zebra-compatible labels suitable for use with Zebra label printers, ensuring consistent, legible slide identification in histology and pathology laboratories.
Specifications
Type Slide Label
Quantity 2500/Roll