Learn More
Electron Microscopy Sciences Plastic 5-Place Side opening Slide Mailer, Natural
SDP
Catalog No. 501929483
Encompass_Preferred
These disposable polypropylene containers are ideal for secure shipping, storing, or staining slides. They hold five standard slides (25 x 75 mm and 1 x 3") and feature slotted channels inside the walls to keep slides separated and for easier removal. The lid closes securely to keep slides safe.
Color: Natural
Catalog No. 50-192-9483 Supplier Electron Microscopy Sciences Supplier No. 7155703
These disposable polypropylene containers are ideal for secure shipping, storing, or staining slides. They hold five standard slides (25 x 75 mm and 1 x 3") and feature slotted channels inside the walls to keep slides separated and for easier removal. The lid closes securely to keep slides safe.
Color: Natural