Learn More
Research Products International Corp Slide Mailer for 2 Slides, Plastic, Natural Color, 25 per Case
SDP
Catalog No. 501978511
Encompass
Mailers offer secure protection for storage and transportation of microscope slides. Each durable polypropylene Slide Mailer holds up to two standard 1 x 3 inch slides securely. Large, easy to open and close snaps secure slides until retreived. Feature large surface for labeling of contents.
Catalog No. 50-197-8511 Supplier Research Products International Corp Supplier No. 212621
Mailers offer secure protection for storage and transportation of microscope slides. Each durable polypropylene Slide Mailer holds up to two standard 1 x 3 inch slides securely. Large, easy to open and close snaps secure slides until retreived. Feature large surface for labeling of contents.