Learn More
Research Products International Corp Slide Mailer for 5 Slides, Plastic, Natural, 25/cs
SDP
Catalog No. 501978508
Encompass
Mailers offer secure protection for storage, transportation or staining of microscope slides. Durable polypropylene container holds up to five standard 1 x 3 inch slides. Slotted interior channels keep slides separated and facilitate quick removal. Lid closes securely to keep slides safe.
Catalog No. 50-197-8508 Supplier Research Products International Corp Supplier No. 212620
Mailers offer secure protection for storage, transportation or staining of microscope slides. Durable polypropylene container holds up to five standard 1 x 3 inch slides. Slotted interior channels keep slides separated and facilitate quick removal. Lid closes securely to keep slides safe.