Learn More
Electron Microscopy Sciences HiQa™ HiQa Slides Mega Twinfrost, Ground Edge
SDP
Catalog No. 501928715
Encompass_Preferred
HiQa Microscope Slides are high quality assured. They have 90 degree ground edges for safety. They are cellophane wrapped, pre-cleaned, ready to use, and suitable for use with automatic coverslipping machines.
Edge: Ground
Catalog No. 50-192-8715 Supplier Electron Microscopy Sciences Supplier No. 7188150
HiQa Microscope Slides are high quality assured. They have 90 degree ground edges for safety. They are cellophane wrapped, pre-cleaned, ready to use, and suitable for use with automatic coverslipping machines.
Edge: Ground