Learn More
IMEB, Inc Tek-Select Microscope Slide Metal Storage Cabinet, 6 Drawers, Stackable
SDP
Catalog No. NC2598170
Encompass_Preferred
Tek-Select Metal Slide Storage Cabinets provide affordable, sturdy slide storage. These cabinets feature convenient pull knob drawers with metal dividers. Removable drawers feature ball suspension design to prevent accidental pull out.Cabinets lock together when stacked, allowing additional levels of stack-able storage on the same amount of counter top or floor space.Cabinet is finished in a powder coat for easy cleaning and durable surface.Each unit has 6 drawers that accommodates up to 5000 slides.
Catalog No. NC2598170 Supplier IMEB, Inc Supplier No. TFD6T
Tek-Select Metal Slide Storage Cabinets provide affordable, sturdy slide storage. These cabinets feature convenient pull knob drawers with metal dividers. Removable drawers feature ball suspension design to prevent accidental pull out.Cabinets lock together when stacked, allowing additional levels of stack-able storage on the same amount of counter top or floor space.Cabinet is finished in a powder coat for easy cleaning and durable surface.Each unit has 6 drawers that accommodates up to 5000 slides.