Learn More
Med Vet International Microscope Slide, Clear, 1in X 3in, 72/Box
Catalog No. 502829838
Encompass_Preferred
High-quality glass slide that will serve every clinical lab need from permanent mounts to fecal samples. Slides individually separated by thin tissue paper to prevent sticking together. Smooth edges with beveled corners1" x 3" x 1.2mm (1/25") thick (clear)
Catalog No. 50-282-9838 Supplier Med Vet International Supplier No. JOR335
High-quality glass slide that will serve every clinical lab need from permanent mounts to fecal samples. Slides individually separated by thin tissue paper to prevent sticking together. Smooth edges with beveled corners1" x 3" x 1.2mm (1/25") thick (clear)