Learn More
Med Vet International Microscope Slide Plain, 72/Box
Catalog No. 502829863
Encompass_Preferred
Pre-cleaned, high quality glass. Hard, colorless, non-corrosive. Ground, polished edges. 1"X3" 72 per box.
Catalog No. 50-282-9863 Supplier Med Vet International Supplier No. MICROPLAIN
Pre-cleaned, high quality glass. Hard, colorless, non-corrosive. Ground, polished edges. 1"X3" 72 per box.