Learn More
Newcomer Supply Slide Moisture Chamber, Clear. 10 slide chamber for keeping your slides well separated, maintaining moisture, and watching the reaction. Stackable, heavy duty plastic construction.
SDP
Catalog No. NC9062082
Encompass
The Slide Staining Tray / Moisture Chamber is perfect for keeping your slides well separated, maintaining moisture, and watching the reaction. The tray is divided into 10 individual compartments with 1/2" empty space between compartments. Eight barrier dividers are placed into empty spaces between compartments when lid is closed for complete isolation. Slides are placed on 4 pedestal posts to raise the surface above the water below. The trays are stackable and made of heavy duty polystyrene. The clear slide staining tray chamber allows easy visualization of the reaction. Dimensions: 8 1/4" x 7" x 1 1/4". Temperature Rating: -10°C to 70°C. Black Slide Staining Tray / Moisture Chamber also available for light sensitive chemistry.
Catalog No. NC9062082 Supplier Newcomer Supply Supplier No. 68431A
The Slide Staining Tray / Moisture Chamber is perfect for keeping your slides well separated, maintaining moisture, and watching the reaction. The tray is divided into 10 individual compartments with 1/2" empty space between compartments. Eight barrier dividers are placed into empty spaces between compartments when lid is closed for complete isolation. Slides are placed on 4 pedestal posts to raise the surface above the water below. The trays are stackable and made of heavy duty polystyrene. The clear slide staining tray chamber allows easy visualization of the reaction. Dimensions: 8 1/4" x 7" x 1 1/4". Temperature Rating: -10°C to 70°C. Black Slide Staining Tray / Moisture Chamber also available for light sensitive chemistry.