Learn More
StatLab Slide Moisture Chamber
SDP
Catalog No. 22130504
Change view
Click to view available options
Color:
Black
Catalog No. Color
22-130-504 Black
1 options
This item is not returnable. View return policy
Catalog No. 22-130-504 Supplier StatLab Supplier No. 197BL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cost effective desktop humidity chamber
  • Maintain humid environments for immunostaining and diagnostic reactions performed on slides
  • Organize work flow with 10 divided compartments with a lid barrier between each slide
  • Ensure climate control with air-tight heavy-duty polystyrene construction
  • Units are stackable. 1 each, unit color is black
Specifications
Type Slide Box
Slide Capacity 10
Color Black