Learn More
Electron Microscopy Sciences Diamond Etch™ Non-Adhesion Slide, White
SDP
Catalog No. 501928577
Encompass_Preferred
Diamond Etch slides, 90 degrees offer optimized etched labeling resolution. Made of white glass designed with top/bottom contrasting colors, specifically for slide etching instruments. Exhibit higher visibility with contrasting background.
Color: White
Catalog No. 50-192-8577 Supplier Electron Microscopy Sciences Supplier No. 7186507
Diamond Etch slides, 90 degrees offer optimized etched labeling resolution. Made of white glass designed with top/bottom contrasting colors, specifically for slide etching instruments. Exhibit higher visibility with contrasting background.
Color: White