Learn More
Research Products International Corp SLIDE OUT FREEZER RACK FOR 2 BOX 12 BOX CAPACITY 3X4 ARRAY 16 1/2 X 5 1/2 X 9 3/4 H
SDP
Catalog No. 50998149
Encompass
Ideal freezer racks feature slide out trays that maximize storage space in upright freezers or refrigerators. Constructed of corrosion resistant stainless steel for rugged durability. Racks will accommodate all 5" square x 2" high cardboard or plastic boxes. Racks are available in several convenient sizes and capacities.
Catalog No. 50-998-149 Supplier Research Products International Corp Supplier No. 247471
Ideal freezer racks feature slide out trays that maximize storage space in upright freezers or refrigerators. Constructed of corrosion resistant stainless steel for rugged durability. Racks will accommodate all 5" square x 2" high cardboard or plastic boxes. Racks are available in several convenient sizes and capacities.